Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium foil, 0.25mm (0.01 in.) thick, hard, 99.8% (metals basis)

Chemical Identifiers

Chemical Name or Material Iridium foil
CAS 7439-88-5
Name Note 0.25mm (0.01 in.) thick, Hard, Temper: as rolled
MDL Number MFCD00011062
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11427FF

  SDS
alfa aesar™
11427FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay Percent Range 99.8% (metals basis)
Physical Form Foil
Weight ≈3.5g/25x25mm
TSCA TSCA
MDL Number MFCD00011062
Quantity 25x25mm
Assay Percent Notes (metals basis)
Name Note 0.25mm (0.01 in.) thick, Hard, Temper: as rolled
Chemical Name or Material Iridium foil
CAS 7439-88-5
Assay 99.8%