missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium black, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 47150ME

 View more versions of this product

Catalog No. AA47150ME



Specifications

Specifications

Iridium black
(metals basis)
MFCD00011062
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
[Ir]
192.217
CHEBI:49666
20 to 30m2/g
500mg
7439-88-5
Ir
UN3089
GKOZUEZYRPOHIO-UHFFFAOYSA-N
iridium
23924
99.95%
Black

Chemical Identifiers

7439-88-5
192.217
GKOZUEZYRPOHIO-UHFFFAOYSA-N
23924
iridium
Ir
MFCD00011062
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
CHEBI:49666
[Ir]
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packaging Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

TSCA : TSCA

SDS Documents