Learn More
Iridium, 99.9%, (trace metal basis), powder, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 198095000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisSpecifications
Gray | |
7439-88-5 | |
Ir | |
500 mg | |
15, 5129 | |
GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
iridium | |
23924 | |
192.2 | |
Glass bottle |
Iridium | |
99.9% | |
MFCD00011062 | |
02,228; 09,257 | |
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge | |
[Ir] | |
192.22 | |
CHEBI:49666 | |
Trace Metal Basis | |
Amorphous Powder |
Chemical Identifiers
7439-88-5 | |
192.22 | |
GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
23924 | |
iridium |
Ir | |
MFCD00011062 | |
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge | |
CHEBI:49666 | |
[Ir] |
Safety and Handling
GHS H Statement:
Flammable solid.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only