Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium, 1% on activated carbon powder, reduced, nominally 50% water wet

$72.18 - $72.18

Chemical Identifiers

Chemical Name or Material Iridium
Name Note 1% on activated carbon powder, Reduced, nominally 50% water wet
MDL Number MFCD00011062
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3833006

  SDS
alfa aesar™
3833006
5g Each for $72.18

AA3833014

  SDS
alfa aesar™
3833014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3833022

  SDS
alfa aesar™
3833022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
TSCA TSCA
MDL Number MFCD00011062
Name Note 1% on activated carbon powder, Reduced, nominally 50% water wet
Chemical Name or Material Iridium