Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium, 0.5% on activated carbon powder, reduced, nominally 50% water wet

Chemical Identifiers

Chemical Name or Material Iridium
Name Note 0.5% on activated carbon powder, Reduced, nominally 50% water wet
MDL Number MFCD00011062
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3832706

 
alfa aesar™
3832706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3832714

 
alfa aesar™
3832714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3832722

 
alfa aesar™
3832722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Iridium
TSCA TSCA
Name Note 0.5% on activated carbon powder, Reduced, nominally 50% water wet
MDL Number MFCD00011062
Quantity 5g