Invitrogen™ IRAK4 Recombinant Human Protein DFS Item
IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases.
Manufacturer:  Invitrogen™ LSPV4002
 View more versions of this product
Catalog No. PV4002
Description
Description
IRAKs are involved in signaling via Toll/IL-1 pathways.
Specifications
Specifications
NP_057207
Frozen
55.6kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
IRAK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
IRAK4
Baculovirus
Human
His Tag
Documents