missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ IRAK4 Recombinant Human Protein DFS Item

IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases.

Manufacturer:  Invitrogen™ LSPV3362

 View more versions of this product

Catalog No. PV3362



Description

Description

IL-1 receptor associated kinases (IRAKs) are serine⁄threonine kinases involved in signaling via Toll⁄IL-1 pathways.
Specifications

Specifications

NP_057207
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Dry Ice
IRAK4
Baculovirus
Full Length, Recombinant
Signal Transduction
Kinase Assay
55.6 kDa
IRAK Kinase Family
Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody)
Ser⁄Thr Kinases: TKL Group
Z'-LYTE™
interleukin-1 receptor-associated kinase 4
Kinases & Inhibitors
Serine⁄Threonine Kinases
Human
51135
IRAK4
His Tag
Documents