missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 56289-64-6 |
---|---|
Molecular Formula | C35H35IN2 |
Molecular Weight (g/mol) | 610.583 |
MDL Number | MFCD07433812 |
InChI Key | PXXBSYXTQAWFHV-UHFFFAOYSA-M |
Synonym | 1,1,3-Trimethyl-2-[5-(1,1,3-trimethyl-1,3-dihydro-2H-benzo[e]indol-2-ylidene)-1,3-pentadienyl]-1H-benzo[e]indolium Iodide |
PubChem CID | 73181418 |
IUPAC Name | 1,1,3-trimethyl-2-[5-(1,1,3-trimethylbenzo[e]indol-3-ium-2-yl)penta-2,4-dienylidene]benzo[e]indole;iodide |
SMILES | CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)C)C=CC=CC=C4C(C5=C(N4C)C=CC6=CC=CC=C65)(C)C)C.[I-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
I1030200MG |
TCI America
I1030200MG |
200mg |
Each for $201.67
|
|
I10301G |
TCI America
I10301G |
1g |
Each for $598.91
|
|
Chemical Identifiers
56289-64-6 | |
610.583 | |
PXXBSYXTQAWFHV-UHFFFAOYSA-M | |
73181418 | |
CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)C)C=CC=CC=C4C(C5=C(N4C)C=CC6=CC=CC=C65)(C)C)C.[I-] |
C35H35IN2 | |
MFCD07433812 | |
1,1,3-Trimethyl-2-[5-(1,1,3-trimethyl-1,3-dihydro-2H-benzo[e]indol-2-ylidene)-1,3-pentadienyl]-1H-benzo[e]indolium Iodide | |
1,1,3-trimethyl-2-[5-(1,1,3-trimethylbenzo[e]indol-3-ium-2-yl)penta-2,4-dienylidene]benzo[e]indole;iodide |
Specifications
Undesignated | |
56289-64-6 | |
MFCD07433812 | |
PXXBSYXTQAWFHV-UHFFFAOYSA-M | |
1,1,3-trimethyl-2-[5-(1,1,3-trimethylbenzo[e]indol-3-ium-2-yl)penta-2,4-dienylidene]benzo[e]indole;iodide | |
73181418 | |
Crystal-Powder at 20°C | |
IR 676 Iodide |
200mg | |
C35H35IN2 | |
1,1,3-Trimethyl-2-[5-(1,1,3-trimethyl-1,3-dihydro-2H-benzo[e]indol-2-ylidene)-1,3-pentadienyl]-1H-benzo[e]indolium Iodide | |
CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)C)C=CC=CC=C4C(C5=C(N4C)C=CC6=CC=CC=C65)(C)C)C.[I-] | |
610.583 | |
610.58 | |
≥97.0% (HPLC,N) |
Safety and Handling
TSCA : No