Please login to your online account to display your discounted pricing

IR-140, Laser Grade 95%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  412565000

 View more versions of this product

Catalog No. AC412565000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Extra Pure
Physical Form Crystals or Crystalline Powder
Formula Weight 778.73
Quantity 500mg
Color Green to dark brown
EINECS Number 258-689-0
TSCA TSCA
Molar Absorption Coefficient 150000L/(mole.cm) (lambda max.)
Packaging Glass bottle
Optimum Lasing 897nm (in DMSO)
Name Note 95%
Fluorescence Max 882nm (in DMSO)
Molecular Formula C39H34Cl3N3O4S2
Infrared Spectrum Authentic
Lambda Max 826nm (in DMSO)
UV_Vis 94% min.
Chemical Name or Material IR-140
Assay Percent Range 95%
MDL Number MFCD00011957
CAS 53655-17-7

Specifications

Chemical Name or Material IR-140
Name Note 95%
CAS 53655-17-7
Purity Grade Extra Pure
Assay Percent Range 95%
Molecular Formula C39H34Cl3N3O4S2
Formula Weight 778.73
MDL Number MFCD00011957