missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IR-140, Laser Grade 95%, ACROS Organics™

Chemical Identifiers

CAS 53655-17-7
Assay Percent Range 95%
Molecular Formula C39H34Cl3N3O4S2
Formula Weight 778.73
MDL Number MFCD00011957
Chemical Name or Material IR-140, 95%
Grade Extra Pure
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC412565000 SDS Thermo Fisher Scientific
412565000
Glass bottle 500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Oxidation reagent
Description & Specifications

Chemical Identifiers

53655-17-7
C39H34Cl3N3O4S2
MFCD00011957
Extra Pure
95%
778.73
IR-140, 95%

Specifications

53655-17-7
95%
C39H34Cl3N3O4S2
MFCD00011957
826nm (in DMSO)
Glass bottle
94% min.
500mg
Extra Pure
Crystals or Crystalline Powder
882nm (in DMSO)
778.73
Authentic
150000L/(mole.cm) (lambda max.)
897nm (in DMSO)
Green to dark brown
IR-140, 95%

Safety and Handling

EINECSNumber : 258-689-0

TSCA : TSCA