Please login to your online account to display your discounted pricing

IQ 1, Tocris™

Enables Wnt/β-catenin-driven expansion of ESCs; prevents spontaneous differentiation

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material IQ 1
Assay Percent Range >99%
Molecular Formula C21H22N4O2
Formula Weight 362.42
Synonym 2-[2-(4-Acetylphenyl)diazenyl]-2-(3, 4-dihydro-3, 3-dimethyl-1(2H)-isoquinolinylidene)acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

471310

 
r & d systems
RDS471310
10mg Each for $225.77

471350

 
r & d systems
RDS471350
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 362.42
Quantity 10mg
Assay Percent Range >99%
Molecular Formula C21H22N4O2
CAS 331001-62-8
Recommended Storage Store at -20°C
Chemical Name or Material IQ 1
Synonym 2-[2-(4-Acetylphenyl)diazenyl]-2-(3, 4-dihydro-3, 3-dimethyl-1(2H)-isoquinolinylidene)acetamide
Safety and Handling

Store at -20°C