Gibco™ iPSC Lab Starter Kit
The Gibco™ iPSC Lab Starter Kit provides the reagents required for researchers initiating pluripotent stem cell research at a significant discount over purchasing these products separately.
Supplier: Gibco™ A32021PKG
Description
This package is appropriate for labs that are starting from cryopreserved pluripotent stem cells. For labs that plan to perform reprogramming of donor-derived fibroblasts or blood cells on feeders and transition to feeder-free culture conditions in the future please refer to A32020 New iPSC Lab Starter Kit with Reprogramming.The kit includes the following products for PSC growth and expansion:
- Essential 8™ Medium
- Geltrex™ hESC-qualified Basement Membrane Extract
- Vitronectin (VTN-N) Substrate
- Versene Solution
- Dulbecco's Phosphate Buffered Saline Without Ca2+/Mg2+
Specifications
Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs), Human Stem Cells | |
Stem Cell Media | |
Dry Ice |