Please login to your online account to display your discounted pricing

IPAG, Tocris


Potent σ antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS054410

 View more versions of this product

Catalog No. 054410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 395.29
CAS 193527-91-2
Chemical Name or Material IPAG
Assay Percent Range >98%
Molecular Formula C17H22IN3
Synonym 1-(4-Iodophenyl)-3-(2-adamantyl)guanidine
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material IPAG
Assay Percent Range >98%
Molecular Formula C17H22IN3
Formula Weight 395.29
Synonym 1-(4-Iodophenyl)-3-(2-adamantyl)guanidine
Accessories

Store at Room Temperature