Please login to your online account to display your discounted pricing

IPAG, Tocris

Potent σ antagonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material IPAG
Assay Percent Range >98%
Molecular Formula C17H22IN3
Formula Weight 395.29
Synonym 1-(4-Iodophenyl)-3-(2-adamantyl)guanidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

054450

 
r & d systems
RDS054450
50mg Each for $746.48

054410

 
r & d systems
RDS054410
10mg Each for $177.37
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 395.29
Assay Percent Range >98%
Molecular Formula C17H22IN3
Chemical Name or Material IPAG
CAS 193527-91-2
Synonym 1-(4-Iodophenyl)-3-(2-adamantyl)guanidine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature