Please login to your online account to display your discounted pricing

Tocris™ IPA 3

Group I p21-activated kinase (PAK) inhibitor

$142.08 - $586.86

Specifications

Product Type IPA 3
Inhibitors IPA 3
Molecular Weight 350.45
Purity >96
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

362210R

 
tocris™
RDS362210
10mg Each for $142.08

362250R

 
tocris™
RDS362250
50mg Each for $586.86
Description & Specifications

Specifications

Product Type IPA 3
Inhibitors IPA 3
Molecular Weight 350.45
Purity >96

Group I p21-activated kinase (PAK) inhibitor (IC50 = 2.5μM at PAK1). Targets the autoregulatory mechanism and promotes the inactive conformation of PAKs. Inhibits PAK1-mediated signaling in vivo; potential anti-tumor agent. Negative control PIR 3.5 (Cat. No. 4212) available.

  • Chemical Name: 1,1'-Dithiodi-2-naphthtol
  • Formula: C20H14O2S2
  • CAS Number: 42521-82-4
  • Solubility: Soluble to 75mM in DMSO and to 10mM in ethanol with gentle warming
  • Purity: >96