Please login to your online account to display your discounted pricing

IOX 2, Tocris™

Potent, selective HIF-1α prolyl hydroxylase-2 (PHD2) inhibitor

$265.15 - $1,116.26

Chemical Identifiers

Chemical Name or Material IOX 2
Assay Percent Range >99%
Molecular Formula C19H16N2O5
Formula Weight 352.34
Synonym N-[[1, 2-Dihydro-4-hydroxy-2-oxo-1-(phenylmethyl)-3-quinolinyl]carbonyl]glycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

445110

 
r & d systems
RDS445110
10mg Each for $265.15

445150

 
r & d systems
RDS445150
50mg Each for $1,116.26
Description & Specifications

Specifications

Formula Weight 352.34
Quantity 10mg
Molecular Formula C19H16N2O5
CAS 931398-72-0
Chemical Name or Material IOX 2
Synonym N-[[1, 2-Dihydro-4-hydroxy-2-oxo-1-(phenylmethyl)-3-quinolinyl]carbonyl]glycine
Assay Percent Range >99%
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C