missing translation for 'onlineSavingsMsg'
Learn More

Iopromide, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018148
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018148 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018148 Supplier Merck Emd Millipore Supplier No. 3870125MG
Only null left

Specifications

CAS 73334-07-3
Physical Form Neat
Linear Formula C18H24I3N3O8
MDL Number MFCD00867924
Type Analytical Standard
Quantity 25 mg
Formula Weight 791.11
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard