Please login to your online account to display your discounted pricing

β-Ionone, 96%, synthetic, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  297135000

 View more versions of this product

Catalog No. AC297135000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid
Formula Weight 192.3
Quantity 500g
Color Clear slightly yellow to yellow
Beilstein 07, 167
Merck Index 10, 4914
RTECS Number EN0350000
TSCA TSCA
EINECS Number 238-969-9
Specific Gravity 0.941 to 0.947 (25°C)
Density 0.9440g/mL
CAS Min % 95.0
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym 4-(2, 6, 6-Trimethyl-1-cyclohexen-1-yl)-3-buten-2-one
Refractive Index 1.5190 to 1.5215 (20°C, 589nm)
Packaging Glass bottle
CAS 14901-07-6
Name Note 96%
Assay Percent Range 96%
MDL Number MFCD00001549
Molecular Formula C13H20O
Chemical Name or Material β-Ionone

Specifications

Chemical Name or Material β-Ionone
Name Note 96%
CAS 14901-07-6
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C13H20O
Formula Weight 192.3
MDL Number MFCD00001549
Synonym 4-(2, 6, 6-Trimethyl-1-cyclohexen-1-yl)-3-buten-2-one