Please login to your online account to display your discounted pricing

Ionomycin (Calcium Salt), Fisher BioReagents


Quantity 5mg
Packaging Poly Micro Tube

Manufacturer: fisher scientific company  BP25275

 View more versions of this product

Catalog No. BP25275

  • / Each
  • / Case of 5 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 747.1
Melting Point 205° to 206°C
Quantity 5mg
Physical Form Solid
Molecular Formula C41H70O9Ca
Color White
Assay Percent Range ≥98.0 %
Purity 98.0% min. (by NP TLC), 99.0% min. (by RP HPLC)
Identification Pass Test
Recommended Storage 2 to 8°C, protect from light
CAS 56092-82-1
ChemAlert Storage Symbol Gray
Name Note From Streptomyces conglobatus
Packaging Poly Micro Tube
Chemical Name or Material Ionomycin, Calcium Salt

Specifications

Chemical Name or Material Ionomycin, Calcium Salt
Name Note From Streptomyces conglobatus
CAS 56092-82-1
Assay Percent Range ≥98.0 %
Molecular Formula C41H70O9Ca
Formula Weight 747.1

  • Ionomycin functions as a calcium ionophore or ion carrier and is commonly used to study Ca2+ transport across biological membranes
  • Ionomycin, in conjunction with PMA, can also be used to induce cytokine production in many cell types
  • Store product at 2-8°C and protected from light
  • Intended for research use only

Accessories
 

2 to 8°C, protect from light

SDS & Safety Documents