Please login to your online account to display your discounted pricing

Alfa Aesar™ Ionomycin calcium salt, 1mg/mL in methanol, sterile-filtered


Quantity 1mL

Manufacturer: alfa aesar™  J67343XF

 View more versions of this product

Catalog No. AAJ67343XF

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

MDL Number MFCD00151183
Health Hazard 2 GHS H Statement
H225-H311-H331-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Ionomycin calcium salt
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL
UN Number UN1230
Health Hazard 1 Danger
Name Note 1mg/mL in methanol, sterile-filtered
Storage Note 1 -30° to -10°C
RTECS Number NO0650000
Physical Form Liquid
Quantity 1mL
CAS 56092-82-1

Specifications

Chemical Name or Material Ionomycin calcium salt
CAS 56092-82-1
Name Note 1mg/mL in methanol, sterile-filtered
MDL Number MFCD00151183
UN Number UN1230
Accessories

Danger

GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H311-H331-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.

-30° to -10°C

SDS & Safety Documents