missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ion exchanger V, EMD-Millipore

Mixed bed exchanger

$506.20 - $3,958.94

Specifications

Chemical Name or Material Ion exchanger V
Synonym Mixed bed exchanger
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M1048360500
View Documents
Emd Millipore Corporation
1048360500
500g Each for $506.20
 
M1048365000
View Documents
Emd Millipore Corporation
1048365000
5kg Each for $3,958.94
 
Specifications

Specifications

Ion exchanger V
Mixed bed exchanger