Please login to your online account to display your discounted pricing

Ion exchanger IV, H+ form, EMD-Millipore

Weakly acidic cation exchanger

$523.46 - $3,487.89

Chemical Identifiers

Chemical Name or Material Ion exchanger IV
Synonym Cation exchangers
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

M1048350500

 
emd millipore corporation
1048350500
500g Each for $523.46

M1048355000

 
emd millipore corporation
1048355000
5kg Each for $3,487.89
Description & Specifications

Specifications

Quantity 500g
Bulk Density 650 to 700kg/m3
Chemical Name or Material Ion exchanger IV
Synonym Cation exchangers
Recommended Storage Store without restrictions
Safety and Handling

Store without restrictions