Please login to your online account to display your discounted pricing

Ion exchanger III, OH- form, EMD-Millipore

Strongly basic anion exchanger

$468.26 - $4,232.49

Chemical Identifiers

Chemical Name or Material Ion exchanger III
Synonym Anion exchangers
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

M1047670500

 
emd millipore corporation
1047670500
500g Each for $468.26

M1047675000

 
emd millipore corporation
1047675000
5kg Each for $4,232.49
Description & Specifications

Specifications

Quantity 500g
Bulk Density 650 to 700kg/m3
Chemical Name or Material Ion exchanger III
Synonym Anion exchangers
Recommended Storage Store without restrictions
Safety and Handling

Store without restrictions