Please login to your online account to display your discounted pricing

Ion exchanger II, OH- form, EMD-Millipore


Weakly basic anion exchanger

Quantity 500mL
Packaging Plastic

Manufacturer: emd millipore corporation  1047680500

 View more versions of this product

Catalog No. M1047680500

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
Density Density: 0.690g/cm3 (20°)
Chemical Name or Material Ion exchanger II
Packaging Plastic
Recommended Storage Store at 15° to 25°C
CAS 63182-01-4

Specifications

Chemical Name or Material Ion exchanger II
CAS 63182-01-4
Accessories

Store at 15° to 25°C