Please login to your online account to display your discounted pricing

Ion exchanger II, OH- form, EMD-Millipore

Weakly basic anion exchanger

$308.40 - $1,860.60

Chemical Identifiers

Chemical Name or Material Ion exchanger II
CAS 63182-01-4
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1047680500

 
emd millipore corporation
1047680500
Plastic 500mL Each for $308.40

M1047685000

 
emd millipore corporation
1047685000
Plastic 5mL Each for $1,860.60
Description & Specifications

Specifications

Quantity 500mL
Density Density: 0.690g/cm3 (20°)
Chemical Name or Material Ion exchanger II
Packaging Plastic
Recommended Storage Store at 15° to 25°C
CAS 63182-01-4
Safety and Handling

Store at 15° to 25°C