Please login to your online account to display your discounted pricing

Ion exchanger II, OH- form, EMD-Millipore


Weakly basic anion exchanger

Quantity 5mL
Packaging Plastic

Manufacturer: emd millipore corporation  1047685000

 View more versions of this product

Catalog No. M1047685000

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 5mL
Density Density: 0.690g/cm3 (20°)
Packaging Plastic
CAS 63182-01-4
Chemical Name or Material Ion exchanger II
Recommended Storage Store at 15° to 25°C

Specifications

Chemical Name or Material Ion exchanger II
CAS 63182-01-4
Accessories

Store at 15° to 25°C