missing translation for 'onlineSavingsMsg'
Learn More

Ion exchanger I, H+ form, Reag. Ph Eur, EMD-Millipore

Strongly acidic cation exchanger

$322.39 - $322.39

Specifications

Chemical Name or Material Ion exchanger I
Density 850kg/m3
Grade Reagent Ph. Eur.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M1047650500
View Documents
Technidata
1047650500
500 g
Each for $322.39
Only null left
 
Description

Description

Specifications

Specifications

Ion exchanger I
850kg/m3
Reagent Ph. Eur.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'