missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Standard Single Element Ion Standard - Cation, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Manufacturer:  Spex Certiprep Llc CSNA22Y

Catalog No. CS-NA2-2Y



Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Na1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Sodium Cation Standard
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
0.1,0.2,99.7
Na
[Na]
22.99
CHEBI:26708
With Certificate of Analysis.
125mL
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
7440-23-5,7732-18-5,7732-18-5
0.1,0.2,99.7
KEAYESYHFKHZAL-UHFFFAOYSA-N
sodium
5360545
Triple-leached LDPE bottle
0.2% HNO3

Chemical Identifiers

7440-23-5
22.99
5360545
sodium
Na
KEAYESYHFKHZAL-UHFFFAOYSA-N
CHEBI:26708
[Na]
Safety and Handling

Safety and Handling

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents
Product Certifications

Product intended for laboratory use only