Please login to your online account to display your discounted pricing

Ion Chromatography Fluoride Standard, 100μg/mL F-, SPEX CertiPrep™


Quantity 500mL
Packaging Poly Bottle

Manufacturer: spex certiprep llc  ASF91X

 View more versions of this product

Catalog No. AS-F9-1X

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
DOT Information Nonhazardous
Recommended Storage Ambient
Solvent or Matrix H2O
Certificate of Analysis With Certificate of Analysis.
Chemical Name or Material Ion Chromatography Fluoride Standard
Concentration or Composition (by Analyte or Components) 100μg/mL F−
CAS Min % 99.99
CAS Max % 0.01
CAS F-
CAS 7732-18-5
CAS Min % 0.01
CAS Max % 99.99
Packaging Poly Bottle

Specifications

Chemical Name or Material Ion Chromatography Fluoride Standard
CAS F-
CAS Min % 0.01
CAS Max % 0.01
CAS 7732-18-5
CAS Min % 99.99
CAS Max % 99.99
Accessories

Ambient