Please login to your online account to display your discounted pricing

Iodotrimethylsilane, 95-97%, stabilized, ACROS Organics™


Quantity 250mL
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC187492500

 View more versions of this product

Catalog No. AC187492500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥95%
Formula Weight 200.1
Color Clear colorless to reddish
Quantity 250mL
Packaging Glass Bottle
Density 1.4000g/mL
UN Number 2924
Boiling Point 106.0°C
Flash Point −31°C
EINECS Number 240-171-0
TSCA TSCA
Fieser 08,261; 09,251; 10,216; 11,271; 12,259; 13,329; 16,188; 06,81
Infrared Spectrum Authentic
Refractive Index 1.4700 to 1.4800 (20°C, 589nm)
Stabilizer copper
Synonym Trimethylsilyl iodide
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Iodotrimethylsilane
Name Note 95-97%
Assay Percent Range 95-97%
Molecular Formula C3H9ISi
Linear Formula (CH3)3SiI
MDL Number MFCD00001028
CAS 16029-98-4

Specifications

Chemical Name or Material Iodotrimethylsilane
Name Note 95-97%
CAS 16029-98-4
Assay ≥95%
Assay Percent Range 95-97%
Molecular Formula C3H9ISi
Linear Formula (CH3)3SiI
Formula Weight 200.1
MDL Number MFCD00001028
Synonym Trimethylsilyl iodide
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.

SDS & Safety Documents