Learn More
Iodosobenzene diacetate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2453136
Description
Iodosobenzene is used as an oxidizing and acetoxylating agent in organic synthesis. It is actively involved in the preparation of (Z)-3,7-dimethyl-2,6-octadien-1-al(neral) from (Z)-3,7-dimethyl-2,6-octadien-1-ol (nerol) in presence of 2,2,6,6-tetramethylpiperidin-1-oxyl (TEMPO). It is a useful reagent for the synthesis of a wide variety of heterocyclic compounds. It is also used in the Pd-catalyzed 2-arylation of indoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIodosobenzene is used as an oxidizing and acetoxylating agent in organic synthesis. It is actively involved in the preparation of (Z)-3,7-dimethyl-2,6-octadien-1-al(neral) from (Z)-3,7-dimethyl-2,6-octadien-1-ol (nerol) in presence of 2,2,6,6-tetramethylpiperidin-1-oxyl (TEMPO). It is a useful reagent for the synthesis of a wide variety of heterocyclic compounds. It is also used in the Pd-catalyzed 2-arylation of indoles.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Specifications
Iodosobenzene diacetate | |
159°C to 161°C | |
MFCD00008692 | |
1879369 | |
iodobenzene diacetate, diacetoxyiodo benzene, iodosobenzene diacetate, phenyliodine diacetate, phenyliodo diacetate, phenyliodoso acetate, phenyliodosodiacetate, phenyliodoso diacetate, bis acetato phenyliodine, benzene, diacetoxyiodo | |
ZBIKORITPGTTGI-UHFFFAOYSA-N | |
[acetyloxy(phenyl)-$l^{3}-iodanyl] acetate | |
76724 | |
≥98% |
3240-34-4 | |
C10H11IO4 | |
500 g | |
Light sensitive | |
Insoluble in water. | |
CC(=O)OI(C1=CC=CC=C1)OC(=O)C | |
322.098 | |
322.1 |
Chemical Identifiers
3240-34-4 | |
322.098 | |
ZBIKORITPGTTGI-UHFFFAOYSA-N | |
76724 | |
CC(=O)OI(C1=CC=CC=C1)OC(=O)C |
C10H11IO4 | |
MFCD00008692 | |
iodobenzene diacetate, diacetoxyiodo benzene, iodosobenzene diacetate, phenyliodine diacetate, phenyliodo diacetate, phenyliodoso acetate, phenyliodosodiacetate, phenyliodoso diacetate, bis acetato phenyliodine, benzene, diacetoxyiodo | |
[acetyloxy(phenyl)-$l^{3}-iodanyl] acetate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H335
EINECSNumber : 221-808-1
RTECSNumber : DA3525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only