Please login to your online account to display your discounted pricing

Iodophenpropit dihydrobromide, Tocris


Potent, selective H3 antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS077910

 View more versions of this product

Catalog No. 077910

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 576.13
Chemical Name or Material Iodophenpropit dihydrobromide
Molecular Formula C15H19IN4S·2HBr
CAS 145196-87-8
Assay Percent Range >98%
Synonym N-[2-(4-Iodophenyl)ethyl]-S-[3-(4(5)-imidazolyl)propyl]isothiourea dihydrobromide
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material Iodophenpropit dihydrobromide
Assay Percent Range >98%
Molecular Formula C15H19IN4S·2HBr
Formula Weight 576.13
Synonym N-[2-(4-Iodophenyl)ethyl]-S-[3-(4(5)-imidazolyl)propyl]isothiourea dihydrobromide
Accessories

Desiccate at +4°C