missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodophenpropit dihydrobromide, Tocris

Potent, selective H3 antagonist

$212.74 - $886.68

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C15H19IN4S·2HBr
Formula Weight 576.13
Synonym N-[2-(4-Iodophenyl)ethyl]-S-[3-(4(5)-imidazolyl)propyl]isothiourea dihydrobromide
Chemical Name or Material Iodophenpropit dihydrobromide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
077950 R & D Systems
RDS077950
50mg Each for $886.68
077910 R & D Systems
RDS077910
10mg Each for $212.74
Specifications Description & Specifications

Chemical Identifiers

>98%
576.13
Iodophenpropit dihydrobromide
C15H19IN4S·2HBr
N-[2-(4-Iodophenyl)ethyl]-S-[3-(4(5)-imidazolyl)propyl]isothiourea dihydrobromide

Specifications

145196-87-8
C15H19IN4S·2HBr
N-[2-(4-Iodophenyl)ethyl]-S-[3-(4(5)-imidazolyl)propyl]isothiourea dihydrobromide
Iodophenpropit dihydrobromide
>98%
576.13
50mg

Safety and Handling

Recommended Storage : Desiccate at +4°C