Please login to your online account to display your discounted pricing

Iodomethane, 99+%, stab. with copper, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Iodomethane
CAS 74-88-4
Name Note stab. with copper
Molecular Formula CH3I
Formula Weight 141.94
MDL Number MFCD00001073
UN Number UN2644
Beilstein 969135
Merck Index 14,6087
Synonym Methyl iodide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA43681A1

  SDS
alfa aesar
43681A1
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 141.94
Quantity 1kg
Color Colorless to Pale Yellow
CAS 74-88-4
Chemical Name or Material Iodomethane
Molecular Formula CH3I
Storage Note 1 2° to 8°C
Merck Index 14,6087
Sensitivity Light Sensitive
Synonym Methyl iodide
Boiling Point 41° to 43°C
UN Number UN2644
TSCA TSCA
Health Hazard 2 GHS H Statement
H301-H331-H351-H312-H315-H335;
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Beilstein 969135
Assay ≥99%
Health Hazard 1 Danger
MDL Number MFCD00001073
Melting Point −67°C
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL IODIDE
Physical Form Liquid
RTECS Number PA9450000
Odor Pungent
Health Hazard 3 GHS P Statement
P261-P280-P281-P301+P310-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note stab. with copper
Safety and Handling

Danger

GHS P Statement
P261-P280-P281-P301+P310-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H351-H312-H315-H335;
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

2° to 8°C