missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodomethane, >99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 4368118*

 View more versions of this product

Catalog No. AA4368118



Specifications

Specifications

Iodomethane
50g
>99%
MFCD00001073
INQOMBQAUSQDDS-UHFFFAOYSA-N
iodomethane
6328
141.94
2.28
74-88-4
CH3I
iodometano
CI
141.939
CHEBI:39282
Liquid

Chemical Identifiers

74-88-4
141.939
INQOMBQAUSQDDS-UHFFFAOYSA-N
6328
iodomethane
CH3I
MFCD00001073
iodometano
CHEBI:39282
CI
Safety and Handling

Safety and Handling

EINECSNumber : 200-819-5

Documents