Please login to your online account to display your discounted pricing

Iodomethane, 98% min., EMD Millipore

$274.83 - $782.17

Chemical Identifiers

Chemical Name or Material Iodomethane
Name Note Stabilized With Silver, For Synthesis
CAS 74-88-4
Physical Form Liquid
Assay Percent Range 98% min.
Molecular Formula CH3I
Linear Formula CH3I
Formula Weight 141.94
Synonym Methyl Iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MIX01851

 
emd millipore corporation
IX01851
100g Each for $274.83

MIX01853

 
emd millipore corporation
IX01853
500g Each for $782.17
Description & Specifications

Specifications

Formula Weight 141.94
Quantity 100g
Color Clear
Physical Form Liquid
Chemical Name or Material Iodomethane
Name Note Stabilized With Silver, For Synthesis
Assay Percent Range 98% min.
Molecular Formula CH3I
Linear Formula CH3I
CAS 74-88-4
Synonym Methyl Iodide
Recommended Storage Store at +15°C to +25°C
Safety and Handling

Store at +15°C to +25°C