missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iodomethane, 98% min., MilliporeSigma™
Supplier: Technidata IX01853
Specifications
Iodomethane | |
74-88-4 | |
CH3I | |
MFCD00001073 | |
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek | |
CI | |
141.94 | |
CHEBI:39282 | |
Liquid |
Stabilized With Silver, For Synthesis | |
98% min. | |
CH3I | |
500 g | |
INQOMBQAUSQDDS-UHFFFAOYSA-N | |
iodomethane | |
6328 | |
141.94 |
Chemical Identifiers
74-88-4 | |
141.94 | |
INQOMBQAUSQDDS-UHFFFAOYSA-N | |
6328 | |
iodomethane |
CH3I | |
MFCD00001073 | |
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek | |
CHEBI:39282 | |
CI |
Safety and Handling
Recommended Storage : Store at +15°C to +25°C