Learn More
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 332121000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
Iodoethane | |
75-03-6,64-19-7 | |
Colorless to Yellow or Pink | |
69.0°C to 73.0°C | |
Authentic | |
Glass bottle | |
1.5100 to 1.5130 | |
100 g | |
1.95 | |
mercury 2+ ion acetic acid | |
HVTICUPFWKNHNG-UHFFFAOYSA-N | |
mercury(2+);diacetate | |
silver | |
0.592 mPa.s (20°C) | |
155.97 | |
Pure |
Stabilized | |
-108.0°C | |
1.9500g/mL | |
62°C | |
97.5% min. (GC) | |
C2H5I | |
MFCD00001091 | |
01,96 | |
15,3868 | |
Solubility in water: 4g/L (20°C),slowly decomposes in water. Other solubilities: miscible with most other organic solvents,soluble in alcohol,ether,chloroform | |
CCI | |
155.97 | |
15337 | |
CHEBI:33211 | |
98% | |
Liquid |
Chemical Identifiers
75-03-6 | |
155.97 | |
HVTICUPFWKNHNG-UHFFFAOYSA-N | |
15337 | |
CCI |
C2H5I | |
MFCD00001091 | |
mercury 2+ ion acetic acid | |
CHEBI:33211 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing dif
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
GHS Signal Word: Danger
EINECSNumber : 200-833-1
RTECSNumber : KI4750000
TSCA : TSCA
Recommended Storage : Product darkens on exposure to light
RUO – Research Use Only