missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iodobenzene, 98%, ACROS Organics™
Manufacturer: Fisher Scientific Acros 122331000
Specifications
Iodobenzene | |
74°C | |
Glass bottle | |
1.82 | |
Yellow | |
100mL | |
97.5% min. (GC) | |
C6H5I | |
01,505 | |
1-iodobenzene, benzene iodide, benzene, iodo, iodinebenzol, iodo benzene, iodo-benzene, iodophenyl, phenyl iodide, phenyliodide, unii-9hk5l7ybbr | |
SNHMUERNLJLMHN-UHFFFAOYSA-N | |
iodobenzene | |
11575 | |
Liquid |
1.8200g/mL | |
Authentic | |
1.6180 to 1.6200 | |
188.0°C | |
-29.0°C | |
591-50-4 | |
C6H5I | |
,215 | |
15,5072 | |
Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether | |
C1=CC=C(C=C1)I | |
204.01 | |
204.01 | |
98% |
Chemical Identifiers
591-50-4 | |
204.01 | |
1-iodobenzene, benzene iodide, benzene, iodo, iodinebenzol, iodo benzene, iodo-benzene, iodophenyl, phenyl iodide, phenyliodide, unii-9hk5l7ybbr | |
iodobenzene |
C6H5I | |
SNHMUERNLJLMHN-UHFFFAOYSA-N | |
11575 | |
C1=CC=C(C=C1)I |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 209-719-6