missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodobenzene, 98%, ACROS Organics™

$120.98 - $120.98

Chemical Identifiers

CAS 591-50-4
Molecular Formula C6H5I
Molecular Weight (g/mol) 204.01
InChI Key SNHMUERNLJLMHN-UHFFFAOYSA-N
Synonym 1-iodobenzene, benzene iodide, benzene, iodo, iodinebenzol, iodo benzene, iodo-benzene, iodophenyl, phenyl iodide, phenyliodide, unii-9hk5l7ybbr
PubChem CID 11575
IUPAC Name iodobenzene
SMILES C1=CC=C(C=C1)I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC122331000
SDS View Documents
Fisher Scientific Acros
122331000
100mL Glass bottle
Each for $120.98
 
AC122335000
SDS View Documents
Thermo Fisher Scientific
122335000
500mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC122330025
SDS View Documents
Thermo Fisher Scientific
AC122330025
2.5L Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

591-50-4
204.01
1-iodobenzene, benzene iodide, benzene, iodo, iodinebenzol, iodo benzene, iodo-benzene, iodophenyl, phenyl iodide, phenyliodide, unii-9hk5l7ybbr
iodobenzene
C6H5I
SNHMUERNLJLMHN-UHFFFAOYSA-N
11575
C1=CC=C(C=C1)I

Specifications

1.8200g/mL
Authentic
1.6180 to 1.6200
188.0°C
-29.0°C
591-50-4
C6H5I
,215
15,5072
Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether
C1=CC=C(C=C1)I
204.01
204.01
98%
74°C
Glass bottle
1.82
Yellow
100mL
97.5% min. (GC)
C6H5I
01,505
1-iodobenzene, benzene iodide, benzene, iodo, iodinebenzol, iodo benzene, iodo-benzene, iodophenyl, phenyl iodide, phenyliodide, unii-9hk5l7ybbr
SNHMUERNLJLMHN-UHFFFAOYSA-N
iodobenzene
11575
Liquid
Iodobenzene
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 209-719-6

SDS