Please login to your online account to display your discounted pricing

Alfa Aesar™ Iodoantipyrine, 99%

$64.34 - $183.82

Chemical Identifiers

Chemical Name or Material Iodoantipyrine
CAS 129-81-7
Molecular Formula C11H11IN2O
Formula Weight 314.13
MDL Number MFCD00040720
Beilstein 181670
Synonym 2,3-Dimethyl-4-iodo-1-phenyl-3-pyrazolin-5-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1400922

  SDS
alfa aesar™
L1400922
100g Each for $183.82

AAL1400914

  SDS
alfa aesar™
L1400914
25g Each for $64.34
Description & Specifications

Specifications

Formula Weight 314.13
Health Hazard 1 Warning
MDL Number MFCD00040720
CAS 129-81-7
Assay 99%
Chemical Name or Material Iodoantipyrine
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C11H11IN2O
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 2,3-Dimethyl-4-iodo-1-phenyl-3-pyrazolin-5-one
RTECS Number UQ9080000
Quantity 100g
Beilstein 181670
Melting Point 155° to 157°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.