Learn More
Iodoacetamide, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 122275000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Iodoacetamide | |
Authentic | |
Plastic Bottle | |
ICH2CONH2 | |
02,223 | |
iodoacetamide, monoiodoacetamide, surauto, acetamide, 2-iodo, 2-iodo-acetamide, usaf d-1, alpha-iodoacetamide, ccris 7710, unii-zrh8m27s79, n-3-methoxy-4-butoxybenzyl thiobarbituric acid | |
PGLTVOMIXTUURA-UHFFFAOYSA-N | |
2-iodoacetamide | |
3727 | |
98% |
144-48-9 | |
97.5% min. (ex I) (Argentometry) | |
C2H4INO | |
500g | |
01,504 | |
Solubility in water: slightly soluble | |
C(C(=O)N)I | |
184.96 | |
184.96 |
Safety and Handling
GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Toxic if swallowed.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
EINECSNumber : 205-630-1
RUO â Research Use Only