missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iodine(V) oxide, 98%

An oxidizing agent to remove CO from gas

Manufacturer:  Alfa Aesar™ 8768214

 View more versions of this product

Catalog No. AA8768214



Specifications

Specifications

Iodine (V) oxide
I2O5
UN3085
iodine pentoxide, iodopentoxide, iodic anhydride, iodine v oxide, diiodine pentaoxide, unii-upu35c0q0n, upu35c0q0n, iodine oxide, jodpentoxyd, iodine pentaoxide
BIZCJSDBWZTASZ-UHFFFAOYSA-N
iodyl iodate
159402
333.81
98%
Light Sensitive & Hygroscopic
275°C to 350°C (decomposition)
25g
12029-98-0
MFCD00011356
14,5019
Soluble in water and HNO3; Insoluble in absolute alcohol, chloroform, ether, and CS2
O=I(=O)OI(=O)=O
333.804
CHEBI:29914
Crystalline
5.08g/mL
White to Cream
1.1

Chemical Identifiers

12029-98-0
333.804
BIZCJSDBWZTASZ-UHFFFAOYSA-N
159402
iodyl iodate
I2O5
MFCD00011356
iodine pentoxide, iodopentoxide, iodic anhydride, iodine v oxide, diiodine pentaoxide, unii-upu35c0q0n, upu35c0q0n, iodine oxide, jodpentoxyd, iodine pentaoxide
CHEBI:29914
O=I(=O)OI(=O)=O
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.

TSCA : TSCA

SDS Documents