missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodine, Sublimated, ACS,ISO,Reag. Ph Eur, For Analysis, EMSURE™, MilliporeSigma™

$879.63 - $879.63

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.809
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym diiodine, eranol, iode, iodine, iodine crystals, iodine solution, iodine sublimed, iodio, tincture iodine, vistarin
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name molecular iodine
SMILES II
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1047610500
View Documents
Emd Millipore Corporation
1047610500
500g Glass bottle Each for $879.63
 
Description

Description

For analysis
Specifications

Chemical Identifiers

7553-56-2
253.809
diiodine, eranol, iode, iodine, iodine crystals, iodine solution, iodine sublimed, iodio, tincture iodine, vistarin
CHEBI:17606
II
I2
PNDPGZBMCMUPRI-UHFFFAOYSA-N
807
molecular iodine

Specifications

7553-56-2
UN 3495
diiodine, eranol, iode, iodine, iodine crystals, iodine solution, iodine sublimed, iodio, tincture iodine, vistarin
II
253.809
CHEBI:17606
Solid
Stinging
0.41 hPa at 25°C
Iodine
I2
Dark violet solid
PNDPGZBMCMUPRI-UHFFFAOYSA-N
molecular iodine
807
253.8. (molar mass)
ACS/ISO/Reagent Ph. Eur.
Glass bottle
500g
Safety and Handling

Safety and Handling

H312 + H332: Harmful in contact with skin or if inhaled
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
H372: Causes damage to organs through prolonged or repeated exposure if swallowed.
H400: Very toxic to aquatic life.

DOTInformation : DOT Class: 8 (6.1), Packaging Group: II

EINECSNumber : 231-442-4