missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodine, Sublimated, ACS,ISO,Reag. Ph Eur, For Analysis, EMSURE™, MilliporeSigma™

Manufacturer:  Emd Millipore Corporation 1047610500

 View more versions of this product

Catalog No. M1047610500



Description

Description

For analysis
Specifications

Specifications

Iodine
I2
Dark violet solid
PNDPGZBMCMUPRI-UHFFFAOYSA-N
molecular iodine
807
253.8. (molar mass)
ACS/ISO/Reagent Ph. Eur.
Glass bottle
500g
7553-56-2
UN 3495
diiodine
II
253.809
CHEBI:17606
Solid
Stinging
0.41 hPa at 25°C

Chemical Identifiers

7553-56-2
253.809
diiodine
CHEBI:17606
II
I2
PNDPGZBMCMUPRI-UHFFFAOYSA-N
807
molecular iodine
Safety and Handling

Safety and Handling

H312 + H332: Harmful in contact with skin or if inhaled
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
H372: Causes damage to organs through prolonged or repeated exposure if swallowed.
H400: Very toxic to aquatic life.

DOTInformation : DOT Class: 8 (6.1), Packaging Group: II

EINECSNumber : 231-442-4

Documents