Please login to your online account to display your discounted pricing

Iodine Solution, Wijs, Fisher Chemical


For determination of iodine absorption number in fats. Approximately 0.22N solution in glacial acetic acid

Quantity 500mL
Packaging Amber Glass

Manufacturer: fisher scientific company  SI106500

 View more versions of this product

Catalog No. SI106-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 17°C
Boiling Point 118°C
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Molecular Formula ClI
Physical Form Liquid
Color Amber
pH 2.0
Density 1.06g/cm³
Vapor Pressure 11mmHg at 20°C
UN Number 2920
CAS 7782-50-5
CAS 64-19-7
Identification Pass test
Health Hazard 2 DANGER!
ChemAlert Storage Symbol White
MDL Number MFCD00011354
Concentration 0.215 to 0.225
Packaging Amber Glass
Halogen Ratio 1.00 to 1.20
Chemical Name or Material Iodine Solution, Wijs
Health Hazard 3 Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:2
Instability:0
CAS 7553-56-2

Specifications

Chemical Name or Material Iodine Solution, Wijs
CAS 7553-56-2
CAS 64-19-7
CAS 7782-50-5
Physical Form Liquid
MDL Number MFCD00011354
Accessories
 

Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:2
Instability:0

DANGER!

SDS & Safety Documents