Please login to your online account to display your discounted pricing

Iodine Solution - Dilute Lugol's Solution, 1.8%

$11.50 - $18.25

Chemical Identifiers

Chemical Name or Material Iodine Solution
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S96854

 
fisher scientific
IX0160-500M
Solution 500mL Each for $11.50

S96870

 
fisher scientific
IX0140-10ML
.05M Solution 10mL Each for $18.25
Description & Specifications

Specifications

Quantity 500mL
Color Amber
Certificate of Analysis Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
Packaging Solution
Chemical Name or Material Iodine Solution
Shelf Life 1 year
Safety and Handling

1 year