Learn More
Iodine solution, according to Wijs, 0.1 mol ICl/l, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 270570010
Specifications
Iodine solution | |
ClI | |
0.098 to 0.113M | |
1.0600g/mL | |
chlorine iodide, chloroiodide, iodine chloride, iodine chloride icl, iodine monochloride, iodinemonochloride, iodochlorine, protochlorure d'iode french, unii-0smg5nlu45, wijs' chloride | |
ClI | |
24640 | |
Liquid | |
1.06 | |
Brown | |
1L |
7790-99-0,64-19-7 | |
MFCD00011354 | |
16,182 | |
15, 5060 | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
162.354 | |
162.36 | |
Glass bottle | |
117.8°C | |
17.2°C | |
40°C |
Chemical Identifiers
7790-99-0 | |
162.354 | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
24640 |
ClI | |
MFCD00011354 | |
chlorine iodide, chloroiodide, iodine chloride, iodine chloride icl, iodine monochloride, iodinemonochloride, iodochlorine, protochlorure d'iode french, unii-0smg5nlu45, wijs' chloride | |
ClI |
Safety and Handling
GHS H Statement:
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTRE or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger