missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iodine, resublimed crystals, 99.9985% (metals basis), Thermo Scientific Chemicals, Puratronic™
Supplier: Thermo Scientific Chemicals 01061914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
183°C to 186°C | |
183°C to 186° (sublimation) | |
Black-Blue | |
25 g | |
I2 | |
3495 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
molecular iodine | |
807 | |
253.81 | |
Bluish-black crystals |
Iodine | |
7553-56-2 | |
Solid | |
99.9985% (Metals basis) | |
MFCD00011355 MFCD00164163 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
II | |
253.81 | |
CHEBI:17606 | |
Irritating | |
4.93 g/cm3 at 20°C |
Chemical Identifiers
7553-56-2 | |
253.81 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
807 | |
II |
I2 | |
MFCD00011355 MFCD00164163 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
CHEBI:17606 |
Safety and Handling
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H372
DOTInformation : DOT Class: 8, Packaging Group: III
EINECSNumber : 231-442-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only