Learn More
Iodine, Reagent ACS, 99.8%, Thermo Scientific Chemicals
$489.46 - $489.46
Chemical Identifiers
CAS | 7553-56-2 |
---|---|
Molecular Formula | I2 |
Molecular Weight (g/mol) | 253.81 |
MDL Number | MFCD00011355 MFCD00164163 |
InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
PubChem CID | 807 |
ChEBI | CHEBI:17606 |
SMILES | II |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC423825000
|
Thermo Scientific Chemicals
423825000 |
500 g | Glass Bottle |
Each for $489.46
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
7553-56-2 | |
253.81 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
807 | |
II |
I2 | |
MFCD00011355 MFCD00164163 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
CHEBI:17606 |
Specifications
185°C | |
7553-56-2 | |
Prills | |
99.8+% | |
I2 | |
3495 | |
15, 5057 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
diiodine | |
807 | |
253.81 | |
ACS Reagent | |
0.41hPa at 25°C | |
Iodine, 99.8% |
113°C | |
Gray | |
500 g | |
I2 | |
MFCD00011355 MFCD00164163 | |
01,495; 02,200; 03,159; 04,258; 05,346; 06,293; 07,179; 08,256; 09,248; 10,210; 11,261; 12,235; 13,133; 14,181; 15,172; 16,182; 17,153 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
II | |
253.81 | |
CHEBI:17606 | |
≥99.80% | |
Glass Bottle | |
8.8 (Air = 1.0) |
Safety and Handling
GHS P Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove to fresh and keep confortable for several minutes.
Warning
EINECSNumber : 231-442-4
RTECSNumber : NN1575000
TSCA : TSCA
RUO – Research Use Only